5-Methoxy-2-nitrophenol, 98%
5-Methoxy-2-nitrophenol, 98%
5-Methoxy-2-nitrophenol, 98%
Thermo Scientific Chemicals

5-Methoxy-2-nitrophenol, 98%

CAS: 704-14-3 | C7H7NO4 | 169.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3354400101 gGlass bottle
Catalog number 335440010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS704-14-3
IUPAC Name5-methoxy-2-nitrophenol
Molecular FormulaC7H7NO4
InChI KeyNRTULWPODYLFOJ-UHFFFAOYSA-N
SMILESCOC1=CC=C(C(O)=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Powder or crystals
Infrared spectrumConforms
GC>=97.5 %
Melting point90°C to 94°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only