(+)-Diacetyl-L-tartaric anhydride, 97%
(+)-Diacetyl-L-tartaric anhydride, 97%
(+)-Diacetyl-L-tartaric anhydride, 97%
Thermo Scientific Chemicals

(+)-Diacetyl-L-tartaric anhydride, 97%

CAS: 6283-74-5 | C8H8O7 | 216.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 336040050
Price (USD)
25.65
Special offer
Online exclusive
Ends: 28-Dec-2026
29.70
Save 4.05
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
25.65
Special offer
Online exclusive
Ends: 28-Dec-2026
29.70
Save 4.05
Each
Chemical Identifiers
CAS6283-74-5
IUPAC Name(3R,4R)-4-(acetyloxy)-2,5-dioxooxolan-3-yl acetate
Molecular FormulaC8H8O7
InChI KeyXAKITKDHDMPGPW-IOMOGOHMNA-N
SMILESCC(=O)O[C@@H]1[C@@H](OC(C)=O)C(=O)OC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Specific optical rotation+56° to +62° (20°C, 589 nm) (c=6, acetone)
Appearance (Form)Powder or crystals or needles
Melting point130°C to 135°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only