Creatine phosphate disodium salt tetrahydrate, 99%
Creatine phosphate disodium salt tetrahydrate, 99%
Creatine phosphate disodium salt tetrahydrate, 99%
Thermo Scientific Chemicals

Creatine phosphate disodium salt tetrahydrate, 99%

CAS: 71519-72-7 | C4H8N3Na2O5P | 255.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
2 g
10 g
Packaging:
Glass bottle
Catalog number 337340010
Price (USD)
82.40
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
82.40
Each
Chemical Identifiers
CAS71519-72-7
IUPAC Namedisodium tetrahydrate ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate
Molecular FormulaC4H16N3Na2O9P
InChI KeyHUWYWJSJJDCZRQ-UHFFFAOYSA-L
SMILESO.O.O.O.[Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<20 ppm
Infrared spectrumConforms
Titration with HClO498.5 to 101.5 %
Water21 to 25 % (K.F.)
Solubility(10 % in water) Clear colorless solution
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only