Dichlorodiethylsilane, 97%
Dichlorodiethylsilane, 97%
Dichlorodiethylsilane, 97%
Thermo Scientific Chemicals

Dichlorodiethylsilane, 97%

CAS: 1719-53-5 | C4H10Cl2Si | 157.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 338100050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1719-53-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light brown
Infrared spectrumConforms
Refractive index1.4290 to 1.4320 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only