tert.-Butyl methyl malonate, 95%
tert.-Butyl methyl malonate, 95%
tert.-Butyl methyl malonate, 95%
Thermo Scientific Chemicals

tert.-Butyl methyl malonate, 95%

CAS: 42726-73-8 | C8H14O4 | 174.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 339390050
Price (USD)
-
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS42726-73-8
IUPAC Name1-tert-butyl 3-methyl propanedioate
Molecular FormulaC8H14O4
InChI KeyXPSYZCWYRWHVCC-UHFFFAOYSA-N
SMILESCOC(=O)CC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
AppearanceMay discolor to yellow on storage
Infrared spectrumConforms
GC>=94 %
Refractive index1.4140 to 1.4170 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only