Pierce™ CN/DAB Substrate Kit
Thermo Scientific™

Pierce™ CN/DAB Substrate Kit

Thermo Scientific Pierce CN/DAB Substrate Kit includes substrate and peroxide solutions for combined chloronaphthol- and diaminobenzidine-based detection of horseradish peroxidaseRead more
Have Questions?
Catalog NumberQuantity
34000275 mL
Catalog number 34000
Price (USD)
190.00
Each
Quantity:
275 mL
Request bulk or custom format
Price (USD)
190.00
Each
Thermo Scientific Pierce CN/DAB Substrate Kit includes substrate and peroxide solutions for combined chloronaphthol- and diaminobenzidine-based detection of horseradish peroxidase (HRP) activity in Western blot and tissue staining methods.

This kit combines two popular chromogenic peroxidase substrate compounds (4-CN and DAB) in a single, stable, 10X stock solution. This CN/DAB solution produces an intense dark black precipitate at sites of bound HRP-conjugated antibodies on probed blots and tissue samples. The Pierce CN/DAB Substrate Kit can be used in Western immunoblotting techniques as well as in dot blotting assays. The kit includes the 10X solution of CN/DAB and accompanying 1X Stable Peroxide Substrate Buffer.

Features of the CN/DAB Substrate Kit:

• HRP substrate—for detection of horseradish peroxidase activity on solid media via the combined action of CN (4-chloro-1-naphthol) and DAB (3,3'-diaminobenzidine tetrahydrochloride)
• Chromogenic—no special equipment needed for visualization; yields intense black bands that are easy to photograph
• Easy-to-use kit—substrate stock solution and stable peroxide substrate buffer mean that there's no dry powders to measure and dissolve
• Stable—store refrigerated at 4°C for at least one year

Includes:
• CN/DAB Solution (10X) and Stable Peroxide Substrate Solution (1X)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ConjugateHRP
ReactivityHorseradish Peroxidase
Sufficient For25 Uses at 10 mL Each
Detection MethodColorimetric
FormLiquid
Product LinePierce
Product TypeSubstrate
Quantity275 mL
SubstrateHorseradish Peroxidase
Unit SizeEach
Contents & Storage
Sufficient For: 25 uses at 10 mL each
• CN/DAB (10X), 25 mL
• Stable Peroxide Buffer, 250 mL

Store at 4°C.