1-Step™ Chloronaphthol Substrate Solution
Thermo Scientific™

1-Step™ Chloronaphthol Substrate Solution

Thermo Scientific Pierce 1-Step Chloronaphthol Solution is a solution of 4-chloro-1-naphthol for the chromogenic detection of horseradish peroxidase (HRP) activityRead more
Have Questions?
Catalog NumberQuantity
34012250 mL
Catalog number 34012
Price (USD)
139.00
Each
Quantity:
250 mL
Request bulk or custom format
Price (USD)
139.00
Each
Thermo Scientific Pierce 1-Step Chloronaphthol Solution is a solution of 4-chloro-1-naphthol for the chromogenic detection of horseradish peroxidase (HRP) activity in Western blot and tissue staining methods.

Chloronaphthol (4-CN, MW = 178.6) reacts with HRP to form a distinct blue to blue-purple product to localize and visualize the activity of peroxidase-conjugated antibodies on blots or fixed tissue samples. The HRP substrate is not as sensitive or stable as other chromogenic substrates, such as TMB and DAB. However, the blue-purple precipitate photographs well, and its alcohol-solubility is a property that can be utilized in certain double-staining applications.

Features of Chloronaphthol Substrates:

• HRP substrate—for detection of horseradish peroxidase activity on solid media
• Chromogenic—no special equipment needed for visualization
• Blue-purple—easy to photograph in color or black-and-white
• Package options—choose powder or ready-to-use solution

Highlights of Pierce CN Solution
• Ready-to-use—no ethanol or methanol required to dissolve; no dilution necessary for use
• Complete—does not require buffer or addition of hydrogen peroxide
• Pre-filtered—does not need to be filtered before use like powders or tablets after they are dissolved
• Stable—store refrigerated at 4°C for at least one year

Related Products
Chloronaphthol Substrate Powder (4-chloro-1-naphthol)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ReactivityHorseradish Peroxidase
Detection MethodColorimetric
FormLiquid
Product Line1-Step
Product TypeSubstrate
Quantity250 mL
SubstrateHorseradish Peroxidase
Unit SizeEach
Contents & Storage
Store at 4°C.