Search
View additional product information for 1-Step™ NBT/BCIP plus Suppressor Substrate Solution - FAQs (34070)
3 product FAQs found
The precipitate formed by the combination of NBT (nitro-blue tetrazolium) and BCIP (5-bromo-4-chloro-3'-indolyphosphate p-toluidine salt) is dark purple in color.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Levamisole is an inhibitor of Alkaline Phosphatase from non-calf intestinal sources. It will cut down on background signal from endogenous alkaline phosphatases.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
No; all of our 1-Step Substrates are ready to use. Simply mix the bottle end over end to ensure homogeneity of the solution and it is ready to go.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.