Nalgene™ PETG Serum Vials; with Crimp Finish: Sterile, Shrink-Wrapped Modules
Actual product may vary
Thermo Scientific™

Nalgene™ PETG Serum Vials; with Crimp Finish: Sterile, Shrink-Wrapped Modules

Protect against leakage, breakage and contamination by using PETG serum vials with crimp finish. Lighter in weight than glass, these vials are made for use with stoppers and caps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Closure Size
342030-001010 mL20 mm
342030-00033 mL13 mm
342030-00055 mL20 mm
342030-002020 mL20 mm
Catalog number 342030-0010
Price (USD)
615.19
Online exclusive
647.57
Save 32.38
Case of 1260
Volume (Metric):
10 mL
Closure Size:
20 mm
Price (USD)
615.19
Online exclusive
647.57
Save 32.38
Case of 1260

Protect against leakage, breakage, and contamination. Thermo Scientific™ Nalgene PETG Crimp Finish Serum Vials are lighter in weight than glass, and are made for use with stoppers and caps.

  • PETG
  • Non-cytotoxic
  • Non-pyrogenic
  • Packaged in trayless shrink-wrap modules
  • USP VI-compliant
  • Sterile to 10-6 SAL

Ordering Information:

Serum Vials are packed in trayless shrink-wrapped modules. Stoppers and Caps not included.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Certifications/ComplianceISO 13485:2016
Closure Size20 mm
DescriptionSerum Vials
Diameter (Metric) Inner13 mm
For Use With (Equipment)Stopper, Cap
Height (Metric)50 mm
No. per Case1260
No. per Pack252
Product TypeSerum Vial
Volume (English)1/3 oz.
Volume (Metric) Total10 mL
Closure TypeStoppers and Caps (Not Included)
ColorClear
Diameter (Metric) Outer24 mm
MaterialPETG
Product LineNalgene
Quantity1260 Cases, 252 Pk.
SterilitySterile
Volume (Metric)10 mL
Unit SizeCase of 1260