Nalgene™ PETG Serum Vials with Continuous Thread: Sterile, Shrink-Wrapped Modules
Nalgene™ PETG Serum Vials with Continuous Thread: Sterile, Shrink-Wrapped Modules
Nalgene™ PETG Serum Vials with Continuous Thread: Sterile, Shrink-Wrapped Modules
Nalgene™ PETG Serum Vials with Continuous Thread: Sterile, Shrink-Wrapped Modules
Nalgene™ PETG Serum Vials with Continuous Thread: Sterile, Shrink-Wrapped Modules
Thermo Scientific™

Nalgene™ PETG Serum Vials with Continuous Thread: Sterile, Shrink-Wrapped Modules

Protect against leakage, breakage and contamination with these PETG serum vials. Lighter in weight than glass, they are made for use with lined continuous-thread closures (available separately).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
342032-001010 mL
342032-00055 mL
342032-002020 mL
Catalog number 342032-0010
Price (USD)
594.30
Online exclusive
660.33
Save 66.03
Case of 1260
Volume (Metric):
10 mL
Price (USD)
594.30
Online exclusive
660.33
Save 66.03
Case of 1260

Protect against leakage, breakage, and contamination with Thermo Scientific™ Nalgene PETG Serum Vials. These serum vials are lighter in weight than glass, and are made for use with lined continuous-thread closures (available separately).

  • PETG
  • Non-cytotoxic
  • Non-pyrogenic
  • Packaged in trayless shrink-wrap modules
  • USP VI-compliant
  • Sterile to 10-6 SAL
  • Leakproof† with Nalgene lined continuous thread closures (Cat. No. 342158)

Ordering Information:

Serum Vials are packed in trayless shrink-wrapped modules. Order closures separately (Cat. No. 342158).

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
Certifications/ComplianceISO 13485:2016
Closure Size20-415
DescriptionSerum Vials
Diameter (Metric) Inner11 mm
For Use With (Equipment)Lined Continuous-Thread Closure
Height (Metric)54 mm
No. per Case1260
No. per Pack252
Product TypeSerum Vial
Volume (English)1/3 oz.
Volume (Metric) Total10 mL
Closure TypeLined Continuous-thread Closure (Sold Separately)
ColorClear
Diameter (Metric) Outer24 mm
MaterialPETG
Product LineNalgene
Quantity1260 Cases, 252 Pk.
SterilitySterile
Volume (Metric)10 mL
Unit SizeCase of 1260