Nalgene™ HDPE, Single-Use Sterile Carboy
Actual product may vary
Thermo Scientific™

Nalgene™ HDPE, Single-Use Sterile Carboy

For Use With: Ideal for storing sterile fluids and pharmaceutical/biotech products.; Closure Material: Polypropylene; Color: White; Sterility: Sterile
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호용량(미터법)
342289-007533 L
342289-005020 L
카탈로그 번호 342289-0075
제품 가격(KRW)
-
용량(미터법):
33 L
Eliminate cleaning, prep and set-up costs with ready-to-use Thermo Scientific™ Nalgene™ Single-Use HDPE Carboys. Supplied sterile for one-time use, the high-density polyethylene vessels are ideal for storing sterile fluids and pharmaceutical/biotech products.

Features

  • HDPE carboy with polypropylene closure
  • Sterile
  • Leakproof†

Ordering Information

Attachable stainless-steel handle available (Cat. No. 2229-0001)

Includes

Polypropylene (PP) closure

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

사양
용량(영국식 단위)8.7 gal.
인증/적합성Food-grade, USP Class VI, non-cytotoxic
마개 재질Polypropylene
마개 크기83B
직경(미터법) 내부 넥64 mm
직경(미터법) 외부381 mm
일회용Single-use
용도(애플리케이션)Storage, Transportation ​
높이(미터법) 마개 포함546 mm
포함Molded-In Handles
워크플로우 단계Upstream media/buffer prep and storage. Upstream process solution storage
오토클레이브 가능Not Autoclavable
용량(미터법)33 L
색상White
물질HDPE
제품 유형Carboy
멸균Sterile
Unit SizeEach