3-Methylfuran, 98%
3-Methylfuran, 98%
3-Methylfuran, 98%
Thermo Scientific Chemicals

3-Methylfuran, 98%

CAS: 930-27-8 | C5H6O | 82.1 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3423000101 gGlass bottle
Catalog number 342300010
Price (USD)
287.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
287.00
Each
Chemical Identifiers
CAS930-27-8
IUPAC Name3-methylfuran
Molecular FormulaC5H6O
InChI KeyKJRRQXYWFQKJIP-UHFFFAOYSA-N
SMILESCC1=COC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4310 to 1.4330 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only