Benzophenone oxime, 98%
Benzophenone oxime, 98%
Benzophenone oxime, 98%
Thermo Scientific Chemicals

Benzophenone oxime, 98%

CAS: 574-66-3 | C13H11NO | 197.24 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 342330250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS574-66-3
IUPAC NameN-(diphenylmethylidene)hydroxylamine
Molecular FormulaC13H11NO
InChI KeyDNYZBFWKVMKMRM-UHFFFAOYSA-N
SMILESON=C(C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Appearance (Form)Powder
Appearance (Color)White
Infrared spectrumConforms
Melting point140°C to 144°C
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only