Product Image
Thermo Scientific Chemicals

(+)-Abscisic acid, 98%

CAS: 21293-29-8 | C15H20O4 | 264.32 g/mol
Have Questions?
Catalog NumberQuantity
342405000500 mg
Catalog number 342405000
Price (USD)
-
Quantity:
500 mg
Request bulk or custom format
Chemical Identifiers
CAS21293-29-8
IUPAC Name(2Z,4E)-5-[(1S)-1-hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl]-3-methylpenta-2,4-dienoic acid
Molecular FormulaC15H20O4
InChI KeyJLIDBLDQVAYHNE-YKALOCIXSA-N
SMILESC\C(\C=C\[C@@]1(O)C(C)=CC(=O)CC1(C)C)=C\C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Specific optical rotation+405° to +413° (20°C, 589 nm) (c=1, EtOH)
Infrared spectrumConforms
Melting point158°C to 164°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only