4-Iodobenzyl bromide, 97%
4-Iodobenzyl bromide, 97%
4-Iodobenzyl bromide, 97%
Thermo Scientific Chemicals

4-Iodobenzyl bromide, 97%

CAS: 16004-15-2 | C7H6BrI | 296.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 342460250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16004-15-2
IUPAC Name1-(bromomethyl)-4-iodobenzene
Molecular FormulaC7H6BrI
InChI KeyBQTRMYJYYNQQGK-UHFFFAOYSA-N
SMILESBrCC1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystals
Infrared spectrumConforms
GC>=96.0 %
Melting point77°C to 82°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only