Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.
Search
View additional product information for Nalgene™ PPCO Low-Profile Closures for Micro Packaging Vials: Sterile, Bulk Pack - FAQs (342821-0118, 342821-0116, 342821-0115, 342821-0114, 342821-0112, 342821-0111, 342821-1111, 342821-0110, 342821-1112)
1 product FAQs found