Nalgene™ PPCO Amber High-Profile Closures with Color Coders for Micro Packaging Vials: Sterile, Bulk Pack, blue - FAQs
Nalgene™ PPCO Amber High-Profile Closures with Color Coders for Micro Packaging Vials: Sterile, Bulk Pack, blue - FAQs
View additional product information for Nalgene™ PPCO Amber High Profile Closures with Color Coders for Micro Packaging Vials: Sterile, Bulk Pack - FAQs (342825-0110, 342825-1111, 342825-1110, 342825-0114, 342825-0113, 342825-0112, 342825-0111, 342825-0118, 342825-0117, 342825-0116, 342825-0115, 342825-0119)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.