Tetramethylammonium hydrogensulfate, 99+%, HPLC grade
Tetramethylammonium hydrogensulfate, 99+%, HPLC grade
Tetramethylammonium hydrogensulfate, 99+%, HPLC grade
Thermo Scientific Chemicals

Tetramethylammonium hydrogensulfate, 99+%, HPLC grade

Green features
CAS: 80526-82-5 | C4H13NO4S | 171.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Packaging:
Glass bottle
Catalog number 343830050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS80526-82-5
IUPAC Nametetramethylazanium hydrogen sulfate
Molecular FormulaC4H13NO4S
InChI KeyDWTYPCUOWWOADE-UHFFFAOYSA-M
SMILESC[N+](C)(C)C.OS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Water=<1 % (K.F.)
UVat 220 nm A: =<0.05
Appearance (Color)White to off-white
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography, Phase transfer catalyst
RUO – Research Use Only