Lanthanum(III) trifluoromethanesulfonate, 99%
Lanthanum(III) trifluoromethanesulfonate, 99%
Lanthanum(III) trifluoromethanesulfonate, 99%
Thermo Scientific Chemicals

Lanthanum(III) trifluoromethanesulfonate, 99%

CAS: 52093-26-2 | C3F9LaO9S3 | 586.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 343910050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS52093-26-2
IUPAC Namelanthanum(3+) tritrifluoromethanesulfonate
Molecular FormulaC3F9LaO9S3
InChI KeyWGJJZRVGLPOKQT-UHFFFAOYSA-K
SMILES[La+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder or crystals
Elemental Analysis23.35 to 24.05 % (La)
Appearance (Color)White to light beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only