Titanium(IV) tert-butoxide, 98%
Titanium(IV) tert-butoxide, 98%
Titanium(IV) tert-butoxide, 98%
Thermo Scientific Chemicals

Titanium(IV) tert-butoxide, 98%

CAS: 3087-39-6 | C16H36O4Ti | 340.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 344000050
Price (USD)
82.65
Online exclusive
91.40
Save 8.75
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
82.65
Online exclusive
91.40
Save 8.75
Each
Chemical Identifiers
CAS3087-39-6
IUPAC Nametitanium(4+) tetrakis(2-methylpropan-2-olate)
Molecular FormulaC16H36O4Ti
InChI KeyGRWPYGBKJYICOO-UHFFFAOYSA-N
SMILES[Ti+4].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Assay>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only