4-Chlorophenylboronic acid, 97%
4-Chlorophenylboronic acid, 97%
4-Chlorophenylboronic acid, 97%
Thermo Scientific Chemicals

4-Chlorophenylboronic acid, 97%

CAS: 1679-18-1 | C6H6BClO2 | 156.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 344410050
Price (USD)
58.65
Online exclusive
65.00
Save 6.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
58.65
Online exclusive
65.00
Save 6.35
Each
Chemical Identifiers
CAS1679-18-1
IUPAC Name(4-chlorophenyl)boronic acid
Molecular FormulaC6H6BClO2
InChI KeyCAYQIZIAYYNFCS-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=96.0 %
Infrared spectrumConforms
Appearance (Color)Off-white to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only