Tetraacetylethylenediamine, 90%
Tetraacetylethylenediamine, 90%
Tetraacetylethylenediamine, 90%
Thermo Scientific Chemicals

Tetraacetylethylenediamine, 90%

CAS: 10543-57-4 | C10H16N2O4 | 228.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Catalog number 344592500
Price (USD)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS10543-57-4
IUPAC NameN-acetyl-N-[2-(N-acetylacetamido)ethyl]acetamide
Molecular FormulaC10H16N2O4
InChI KeyBGRWYDHXPHLNKA-UHFFFAOYSA-N
SMILESCC(=O)N(CCN(C(C)=O)C(C)=O)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to beige
Appearance (Form)Granular powder
Infrared spectrumConforms
Assay>=88 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

Is this Tetraacetylethylenediamine (TAED), 90% (Cat. Nos. 344592500, 344590500) uncoated ?

Yes, this Tetraacetylethylenediamine, 90% is uncoated.