2-Chloro-4-aminopyridine, 97%
2-Chloro-4-aminopyridine, 97%
2-Chloro-4-aminopyridine, 97%
Thermo Scientific Chemicals

2-Chloro-4-aminopyridine, 97%

CAS: 14432-12-3 | C5H5ClN2 | 128.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 345450050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14432-12-3
IUPAC Name2-chloropyridin-4-amine
Molecular FormulaC5H5ClN2
InChI KeyBLBDTBCGPHPIJK-UHFFFAOYSA-N
SMILESNC1=CC=NC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HClO4>=96.0 %
Appearance (Color)Light yellow
Infrared spectrumConforms
Melting point89°C to 94°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only