Water, Extra Pure, Deionized
Water, Extra Pure, Deionized
Water, Extra Pure, Deionized
Thermo Scientific Chemicals

Water, Extra Pure, Deionized

CAS: 7732-18-5 | H2O | 18.015 g/mol
製品番号(カタログ番号)数量パッケージ
34547025025 LPlastic Drum
製品番号(カタログ番号) 345470250
価格(JPY)
-
数量:
25 L
パッケージ:
Plastic Drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7732-18-5
IUPAC Namewater
Molecular FormulaH2O
InChI KeyXLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILESO
さらに表示
Appearance (Form)Liquid
Residue after evaporation=<25 ppm
Conductivity=<5.0 µS/cm
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only