1,2,3-1H-Triazole, 97%
1,2,3-1H-Triazole, 97%
1,2,3-1H-Triazole, 97%
Thermo Scientific Chemicals

1,2,3-1H-Triazole, 97%

CAS: 288-36-8 | C2H3N3 | 69.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 345480010
Price (USD)
49.65
Online exclusive
55.10
Save 5.45
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
49.65
Online exclusive
55.10
Save 5.45
Each
Chemical Identifiers
CAS288-36-8
IUPAC Name2H-1,2,3-triazole
Molecular FormulaC2H3N3
InChI KeyQWENRTYMTSOGBR-UHFFFAOYSA-N
SMILESN1N=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
GC>=96.0 %
Appearance (Form)Liquid
Refractive index1.4940 to 1.4980 (20°C, 589 nm)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only