4-(tert-Butyl)benzylbromide, 97%
4-(tert-Butyl)benzylbromide, 97%
4-(tert-Butyl)benzylbromide, 97%
Thermo Scientific Chemicals

4-(tert-Butyl)benzylbromide, 97%

CAS: 18880-00-7 | C11H15Br | 227.14 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
產品號碼 345500250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS18880-00-7
IUPAC Name1-(bromomethyl)-4-tert-butylbenzene
Molecular FormulaC11H15Br
InChI KeyQZNQSIHCDAGZIA-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(CBr)C=C1
檢視更多
規格Specification Sheet規格表
Appearance (Form)Liquid
Titration Argentometric>=96.0 %
Infrared spectrumConforms
Refractive index1.5445 to 1.5465 (20°C, 589 nm)
Appearance (Color)Clear light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only