3-Fluorophenylboronic acid, 97%
3-Fluorophenylboronic acid, 97%
3-Fluorophenylboronic acid, 97%
Thermo Scientific Chemicals

3-Fluorophenylboronic acid, 97%

CAS: 768-35-4 | C6H6BFO2 | 139.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 345640010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS768-35-4
IUPAC Name(3-fluorophenyl)boronic acid
Molecular FormulaC6H6BFO2
InChI KeyKNXQDJCZSVHEIW-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
Titration with NaOH>=96.0 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only