SuperSignal™ West Pico PLUS Chemiluminescent Substrate, 4 x 1 L - FAQs
View additional product information for SuperSignal™ West Pico PLUS Chemiluminescent Substrate - FAQs (34579, 34578, 34577, 34580X4, 34580, 34578X4)
1 product FAQs found
How do I make the working solution for the SuperSignal West Pico PLUS Chemiluminescent Substrate?
To prepare the working solution for the SuperSignal West Pico PLUS Chemiluminescent Substrate, follow these steps:
- Mix equal parts of the SuperSignal West Pico PLUS Luminol/Enhancer Solution and SuperSignal West Pico PLUS Stable Peroxide Solution. For example, for a mini blot, mix 5 mL of SuperSignal West Pico PLUS Luminol/Enhancer Solution with 5 mL of SuperSignal West Pico PLUS Stable Peroxide Solution.
- Ensure that the volume is sufficient to completely wet the blot and prevent it from drying out. The recommended volume is 0.1 mL/cm2.
- Note that the working solution is stable for up to 8 hours at room temperature. Avoid exposure to the sun or other intense light. Short-term exposure to lab lighting is okay.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.