PCR Ready Knurl Microcentrifuge Tube, Amber
Molecular Bio Products Inc.

PCR Ready Knurl Microcentrifuge Tube, Amber

Protects light-sensitive samples with secure sealing.
Have Questions?
Catalog NumberCapacity (Metric)
3461A1.5 mL, 1.5 mL, 1.5 mL, 1.5 mL
Catalog number 3461A
Price (USD)
-
Capacity (Metric):
1.5 mL, 1.5 mL, 1.5 mL, 1.5 mL
Protects light-sensitive samples with secure sealing.
  • Amber protection
  • 1.5 mL capacity
  • Sterile
  • Secure screw cap with o-ring
  • High RCF tolerance
Specifications
Binding PropertyLow Nucleic Acid and Protein Binding & Low Enzyme Inhibition
Clean ClaimsSterile Electron Beam, SAL 10-6 Free of: Rnase/Dnase, Human and Mouse DNA, Pyrogen, Endotoxin, Bioburden, ATP & PCR Inhibitor
Closure IncludedYes
ColorAmber
IncludesKnurl
Max. TorqueManual/Hand Torque: Standard: 0.125 lbsf.in to 2.5 lbsf.in; Metric: 1.412 Ncm to 28.24 Ncm. Automated/Robotic Torque Standard: 1.312 lbsf.in to 2.5 lbsf.in; Metric: 14.83 Ncm to 28.24 Ncm
Packaging500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
Product TypeScrew-cap Microtube
Quantity500 Pk.
Temperature Range (Metric)Cold Storage: -80° to +95°C. Liquid Nitrogen: Short term Vapor Phase & Quick Flash Freeze acceptable, not recommmended for LN2 submersion storage
UV Resistance(UV) Ultra violet protection avg. 1.22% transmittance between 220 and 500nm
AutoclavableNot Autoclavable, Not Autoclavable, Not Autoclavable, Not Autoclavable
Bottom ShapeNon-skirted, Non-skirted, Non-skirted, Non-skirted
Capacity (Metric)1.5 mL, 1.5 mL, 1.5 mL, 1.5 mL
Closure TypeAttached Cap, Attached Screw Cap, Attached Cap, Attached Screw Cap
ColorAmber
Fit for PurposeSterile and PCR Ready
GraduatedYes
MaterialPolypropylene, Polypropylene (Virgin), Polypropylene, Polypropylene (Virgin)
Max. RCF20,000 x g, 18,000 x g, 20,000 x g, 18,000 x g
SterilitySterile, Sterile, Sterile, Sterile
Unit SizeCase of 5000