trans-4-Aminocyclohexanol, 97%
trans-4-Aminocyclohexanol, 97%
trans-4-Aminocyclohexanol, 97%
Thermo Scientific Chemicals

trans-4-Aminocyclohexanol, 97%

CAS: 27489-62-9 | C6H13NO | 115.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number 346680250
Price (EUR)
109,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
109,00
Each
Chemical Identifiers
CAS27489-62-9
IUPAC Name4-aminocyclohexan-1-ol
Molecular FormulaC6H13NO
InChI KeyIMLXLGZJLAOKJN-UHFFFAOYSA-N
SMILESNC1CCC(O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Melting point108°C to 113°C
Infrared spectrumConforms
Appearance (Color)White to cream to light brown
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only