D(-)-Phenylglycinamide, 99%
D(-)-Phenylglycinamide, 99%
D(-)-Phenylglycinamide, 99%
Thermo Scientific Chemicals

D(-)-Phenylglycinamide, 99%

CAS: 6485-67-2 | C8H10N2O | 150.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 346700050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6485-67-2
IUPAC Name(R)-carbamoyl(phenyl)methanaminium
Molecular FormulaC8H11N2O
InChI KeyKIYRSYYOVDHSPG-SSDOTTSWSA-O
SMILESNC(=O)[C@H]([NH3+])C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Specific optical rotation-110° to -120° (20°C, 589 nm) (c=0.55, EtOH)
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only