1-Acetylpiperazine, 98%
1-Acetylpiperazine, 98%
1-Acetylpiperazine, 98%
Thermo Scientific Chemicals

1-Acetylpiperazine, 98%

CAS: 13889-98-0 | C6H12N2O | 128.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 346830250
Price (USD)
147.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
147.00
Each
Chemical Identifiers
CAS13889-98-0
IUPAC Name4-acetylpiperazin-1-ium
Molecular FormulaC6H13N2O
InChI KeyPKDPUENCROCRCH-UHFFFAOYSA-O
SMILESCC(=O)N1CC[NH2+]CC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)Clear light yellow
Appearance (Form)Liquid after melting
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only