Brij™ 98
Brij™ 98
Brij™ 98
Thermo Scientific Chemicals

Brij™ 98

CAS: 9004-98-2 | C58H116O21 | 1149.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 347181000
Price (USD)
34.65
Special offer
Online exclusive
Ends: 28-Dec-2026
40.80
Save 6.15
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
34.65
Special offer
Online exclusive
Ends: 28-Dec-2026
40.80
Save 6.15
Each
Chemical Identifiers
CAS9004-98-2
IUPAC Name(69Z)-3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxaoctaheptacont-69-en-1-ol
InChI KeyTVSFHLHLHUYZNN-KTKRTIGZSA-N
SMILESCCCCCCCC\C=C/CCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO
Synonym(69Z)-3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxaoctaheptacont-69-en-1-ol
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity=<1 ppm free Ethyleneoxide
Antioxidantwithout Antioxidant
Antioxidantwithout Antioxidant
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only