Screw Cap Microcentrifuge Tube Caps
Thermo Scientific™
Screw Cap Microcentrifuge Tube Caps
Prevent leakage under demanding conditions with these screw-cap microcentrifuge tube caps with o-rings, which are optimized for secure, short-term storage in low temperatures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorProduct TypeSterilityFit for PurposeUnit Size
3471YSYellowScrew Cap with O-ringSterileSterile and PCR ReadyCase of 5000
3471AAmberScrew Cap with O-ringNon-sterilePCR ReadyCase of 5000
3471ASAmberScrew Cap with O-ringSterileSterile and PCR ReadyCase of 5000
3471BLKBlackScrew Cap with O-ringNon-sterilePCR ReadyCase of 5000
3471BLKSBlackScrew Cap with O-ringSterileSterile and PCR ReadyCase of 5000
3471BBlueScrew Cap with O-ringNon-sterilePCR ReadyEA
3471BPK
also known as 3471B
BlueScrew Cap with O-ringNon-sterilePCR ReadyPack of 500
3471BSBlueScrew Cap with O-ringSterileSterile and PCR ReadyCase of 5000
3471GGreenScrew Cap with O-ringNon-sterilePCR ReadyCase of 5000
3471GPK
also known as 3471G
GreenScrew Cap with O-ringNon-sterilePCR ReadyPack of 500
3471GSGreenScrew Cap with O-ringSterileSterile and PCR ReadyCase of 5000
3471NaturalScrew Cap with O-ringNon-sterilePCR ReadyCase of 5000
3471PKNaturalScrew Cap with O-ringNon-sterilePCR ReadyPack of 500
3471SNaturalScrew Cap with O-ringSterileSterile and PCR ReadyCase of 5000
3471OOrangeScrew Cap with O-ringNon-sterilePCR ReadyCase of 5000
3471OSOrangeScrew Cap with O-ringSterileSterile and PCR ReadyCase of 5000
3471PPinkScrew Cap with O-ringNon-sterilePCR ReadyCase of 5000
3471PSPinkScrew Cap with O-ringSterileSterile and PCR ReadyCase of 5000
3471RRedScrew Cap with O-ringNon-sterilePCR ReadyCase of 5000
3471RPK
also known as 3471R
RedScrew Cap with O-ringNon-sterilePCR ReadyPack of 500
3471RSRedScrew Cap with O-ringSterileSterile and PCR ReadyCase of 5000
3471VVioletScrew Cap with O-ringNon-sterilePCR ReadyCase of 5000
3471VSVioletScrew Cap with O-ringSterileSterile and PCR ReadyCase of 5000
3471WWhiteScrew Cap with O-ringNon-sterilePCR ReadyCase of 5000
3471WSWhiteScrew Cap with O-ringSterileSterile and PCR ReadyCase of 5000
3471YYellowScrew Cap with O-ringNon-sterilePCR ReadyCase of 5000
3471YPK
also known as 3471Y
YellowScrew Cap with O-ringNon-sterilePCR ReadyPack of 500
3471MIXAssorted (Black, Blue, Green, Natural, Orange, Pink, Red, Violet, White, Yellow)Screw Cap with O-ringNon-sterilePCR ReadyCase of 5000
3471TAAmberTether Screw Cap with O-ringNon-sterilePCR ReadyEach
3471TASAmberTether Screw Cap with O-ringSterileSterile and PCR ReadyEach
3471TBLKBlackTether Screw Cap with O-ringNon-sterilePCR ReadyEach
3471TBLKSBlackTether Screw Cap with O-ringSterileSterile and PCR ReadyEach
3471TBBlueTether Screw Cap with O-ringNon-sterilePCR ReadyEach
3471TBSBlueTether Screw Cap with O-ringSterileSterile and PCR ReadyEach
3471TGGreenTether Screw Cap with O-ringNon-sterilePCR ReadyEach
3471TGSGreenTether Screw Cap with O-ringSterileSterile and PCR ReadyEach
3471TNaturalTether Screw Cap with O-ringNon-sterilePCR ReadyEach
3471TSNaturalTether Screw Cap with O-ringSterileSterile and PCR ReadyEach
3471TOOrangeTether Screw Cap with O-ringNon-sterilePCR ReadyEach
3471TOSOrangeTether Screw Cap with O-ringSterileSterile and PCR ReadyEach
3471TPPinkTether Screw Cap with O-ringNon-sterilePCR ReadyEach
3471TPSPinkTether Screw Cap with O-ringSterileSterile and PCR ReadyEach
3471TRRedTether Screw Cap with O-ringNon-sterilePCR ReadyEach
3471TRSRedTether Screw Cap with O-ringSterileSterile and PCR ReadyEach
3471TVVioletTether Screw Cap with O-ringNon-sterilePCR ReadyEach
3471TVSVioletTether Screw Cap with O-ringSterileSterile and PCR ReadyEach
3471TWWhiteTether Screw Cap with O-ringNon-sterilePCR ReadyEach
3471TWSWhiteTether Screw Cap with O-ringSterileSterile and PCR ReadyEach
3471TYYellowTether Screw Cap with O-ringNon-sterilePCR ReadyEach
3471TYSYellowTether Screw Cap with O-ringSterileSterile and PCR ReadyEach
3471TMIXAssorted (Black, Blue, Green, Natural, Orange, Pink, Red, Violet, White, Yellow)Tether Screw Cap with O-ringPCR ReadyEach
3471TMIXSAssorted (Black, Blue, Green, Natural, Orange, Pink, Red, Violet, White, Yellow)Tether Screw Cap with O-ringSterileSterile and PCR ReadyEach
Catalog number 3471YS
Price (USD)
-
Color:
Yellow
Product Type:
Screw Cap with O-ring
Sterility:
Sterile
Fit for Purpose:
Sterile and PCR Ready
Unit Size:
Case of 5000
Prevent leakage under demanding conditions with Thermo Scientific™ Screw Cap Microcentrifuge Tube Caps feature O-Rings to form a secure seal. Optimized for secure, short-term storage in low temperatures, these non-autoclavable screw caps are available in a variety of colors and adapted to your scientific experiments.
Non-Sterile is free of Rnase/Dnase, Human and Mouse DNA, Pyrogen & Endotoxin, ATP, PCR Inhibitor.
Sterile E-Beam is free of Rnase/Dnase, Human and Mouse DNA, Pyrogen, Endotoxin, ATP, PCR Inhibitor.
100% virgin polypropylene resin specifically selected for high clarity and biologic applications.
O-ring design, thickness and material selection have been optimized to provide protective features for maximum sealing ability, biologic interaction and temperature capability. The black colored O-ring is made from EPDM.
Knurled caps improve grip when opening and closing tube.
Frosted cap writing pad for easy product marking and identification.
Color caps offered in multiple color options ideal for separating, coding and tracking samples. Amber colored caps ideal for UV protection of 1.22% transmittance between 220 and 500nm.
Tethered caps remain attached to the sample tube but can rotate completely for convenience — the tethered screw cap is the solution when it is important to maintain the same cap with the same tube. Non-tethered caps available.
RCF range from 18,000 to 25,000 x g.
Temperature Range -80°C to +95°C. Short term storage in     -80°C, -20°C, dry ice and wet ice. Short term storage in vapor phase LN2 and suitable for a quick dip flash freeze in LN2.
Torque Manual/Hand Torque: Standard: 0.125 lbsf.in to 2.5 lbsf.in; Metric: 1.412 Ncm to 28.24 Ncm.
Torque Automated/Robotic Torque Standard: 1.312 lbsf.in to 2.5 lbsf.in; Metric: 14.83 Ncm to 28.24 Ncm.
Screw cap micro tubes and caps are not recommended for autoclave. Sterile options available.
Not intended to submerge and store in liquid phase LN2.
Specifications
Binding PropertyLow Nucleic Acid and Protein Binding & Low Enzyme Inhibition
Clean ClaimsSterile Electron Beam, SAL 10-6 Free of: Rnase/Dnase, Human and Mouse DNA, Pyrogen, Endotoxin, Bioburden, ATP & PCR Inhibitor
Max. RCF25,000 x g
Max. TorqueManual/Hand Torque: Standard: 0.125 lbsf.in to 2.5 lbsf.in; Metric: 1.412 Ncm to 28.24 Ncm. Automated/Robotic Torque Standard: 1.312 lbsf.in to 2.5 lbsf.in; Metric: 14.83 Ncm to 28.24 Ncm
Packaging500 x pk, 10 pk x cs, 5000 x cs. Single Bagged. ZipLock bag.
Temperature Range (Metric)Cold Storage: -80° to +95°C. Liquid Nitrogen: Short term Vapor Phase & Quick Flash Freeze acceptable, not recommmended for LN2 submersion storage
AutoclavableNot Autoclavable
ColorYellow
Fit for PurposeSterile and PCR Ready
MaterialPolypropylene, Ethylene Propylene
Product TypeScrew Cap with O-ring
SterilitySterile
Unit SizeCase of 5000