2',7'-Dichlorofluorescein 3',6'-diacetate, 97%
2',7'-Dichlorofluorescein 3',6'-diacetate, 97%
2',7'-Dichlorofluorescein 3',6'-diacetate, 97%
Thermo Scientific Chemicals

2',7'-Dichlorofluorescein 3',6'-diacetate, 97%

CAS: 2044-85-1 | C24H14Cl2O7 | 485.27 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3472500101 gGlass bottle
Catalog number 347250010
Price (USD)
122.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
122.00
Each
Chemical Identifiers
CAS2044-85-1
IUPAC Name3'-(acetyloxy)-2',7'-dichloro-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-6'-yl acetate
Molecular FormulaC24H14Cl2O7
InChI KeyVQVUBYASAICPFU-UHFFFAOYSA-N
SMILESCC(=O)OC1=CC2=C(C=C1Cl)C1(OC(=O)C3=CC=CC=C13)C1=CC(Cl)=C(OC(C)=O)C=C1O2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to off-white
Infrared spectrumConforms
Appearance (Form)Powder or crystals
Melting point239°C to 246°C
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only