8-Hydroxyquinoline-2-carboxaldehyde, 98%
8-Hydroxyquinoline-2-carboxaldehyde, 98%
8-Hydroxyquinoline-2-carboxaldehyde, 98%
Thermo Scientific Chemicals

8-Hydroxyquinoline-2-carboxaldehyde, 98%

CAS: 14510-06-6 | C10H7NO2 | 173.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3476300101 gGlass bottle
Catalog number 347630010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14510-06-6
IUPAC Name8-hydroxyquinoline-2-carbaldehyde
Molecular FormulaC10H7NO2
InChI KeySLBPIHCMXPQAIQ-UHFFFAOYSA-N
SMILESOC1=C2N=C(C=O)C=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
GC>=97.5 %
Appearance (Color)Yellow
Infrared spectrumConforms
Melting point96°C to 100°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only