6-Hydroxy-2-naphthoic acid, 98%
6-Hydroxy-2-naphthoic acid, 98%
6-Hydroxy-2-naphthoic acid, 98%
Thermo Scientific Chemicals

6-Hydroxy-2-naphthoic acid, 98%

CAS: 16712-64-4 | C11H8O3 | 188.18 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3476500505 gGlass bottle
Catalog number 347650050
Price (USD)
60.10
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
60.10
Each
Chemical Identifiers
CAS16712-64-4
IUPAC Name6-hydroxynaphthalene-2-carboxylic acid
Molecular FormulaC11H8O3
InChI KeyKAUQJMHLAFIZDU-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C2C=C(O)C=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Titration with TBAH>=97.5 %
Melting point240°C to 250°C
Appearance (Color)Light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only