Nunc™ General Long Term Storage Tubes, Nunc Stor-It Tubes, 1.8mL, 48mm, ext. thread, non-sterile - FAQs
Nunc™ General Long Term Storage Tubes, Nunc Stor-It Tubes, 1.8mL, 48mm, ext. thread, non-sterile - FAQs
View additional product information for Nunc™ General Long Term Storage Tubes - FAQs (347783, 347791)
1 product FAQs found
Do you offer non-sterile Nunc Cryo Tubes?
We offer Nunc General Long Term Storage Tubes that are non-sterile and available in 1.8 mL and 4.5 mL capacities.