Nunc™ 10/11mL Polystyrene Centrifuge Tubes
Actual product may vary
Thermo Scientific™

Nunc™ 10/11mL Polystyrene Centrifuge Tubes

Increase the effectiveness of centrifugation with these polystyrene centrifuge tubes, which have been both RCF and tightness tested.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBottom ShapeGraduationsWriting AreaSterility
348224RoundNoSterile
337846ConicalYesSterile
347856ConicalNoSterile
347880ConicalYesSterile
Catalog number 348224
Price (USD)
-
Bottom Shape:
Round
Writing Area:
No
Sterility:
Sterile
Increase the effectiveness of centrifugation with Thermo Scientific™ Nunc™ 10/11mL Polystyrene Centrifuge Tubes, which have been both RCF and tightness tested.
  • Made in a variety of formats for greater flexibility
  • Different bottom shapes to meet all needs
  • Include polyethylene plug seal screw caps
  • Fit most standard centrifuges
  • Relative centrifugal force tested (RCF)
Specifications
Diameter (Metric)16 mm
Length (Metric)110 mm
No. per Case480
No. per Pack80
Product TypeCentrifuge Tube
ShapeRound
Writing AreaNo
Bottom ShapeRound
Capacity (Metric)10 mL
GraduatedNo
MaterialPolystyrene
Max. RCF3000 x g
SterilitySterile
Unit SizeCase of 480