Succinimide, 98%
Succinimide, 98%
Succinimide, 98%
Thermo Scientific Chemicals

Succinimide, 98%

CAS: 123-56-8 | C4H5NO2 | 99.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 348650050
Price (USD)
40.30
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
40.30
Each
Chemical Identifiers
CAS123-56-8
IUPAC Namepyrrolidine-2,5-dione
Molecular FormulaC4H5NO2
InChI KeyKZNICNPSHKQLFF-UHFFFAOYSA-N
SMILESO=C1CCC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to beige to light brown
Appearance (Form)Powder or flakes
Infrared spectrumConforms
Melting point121°C to 126°C
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only