Microtiter Tubes and Racks
Thermo Scientific™
Microtiter Tubes and Racks
Seal microtiter tubes tightly for storage with these polyethylene plug strips -- for use with 1.2mL MCT titertube racks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeSterilityFormatUnit Size
3487Microtiter TubeSterileRobotic RackCase of 4800
3496PK
also known as 3496
Microtiter TubeNon-sterileRobotic RackPack of 960
3492PK
also known as 3492
Microtiter TubeNon-sterileBulkPack of 1000
3487PK
also known as 3487
Microtiter TubeSterileRobotic RackPack of 960
3492Microtiter TubeNon-sterileBulkCase of 10000
Catalog number 3487
Price (USD)
320.87
Online exclusive
357.00
Save 36.13
Case of 4800
Product Type:
Microtiter Tube
Sterility:
Sterile
Format:
Robotic Rack
Unit Size:
Case of 4800
Price (USD)
320.87
Online exclusive
357.00
Save 36.13
Case of 4800
Seal microtiter tubes tightly for storage with polyethylene Thermo Scientific™ Microtiter Tube Plug Strips — for use with 1.2mL MCT Titertube Racks. The 1.2mL microtiter tubes offer additional volume capacity for sample dilution. These are available in a pre-racked 8 x 12 format.
Microtiter Tubes and Caps
  • Polyethylene plug strip seals the microtiter tubes tightly for storage
  • Eight-plug strips can be easily cut apart for individual samples
  • 1.2mL microtiter tubes have additional volume capacity for sample dilution and are ideal for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control and blood bank sample freezer storage
  • Frosted writing areas on each tube and an alphanumeric rack simplify organization and identification of specific samples
  • Microtiter tube system is available in bulk packaging or with a SBS standard footprint
  • Robotic compatible rack can easily integrated into automated liquid handling workstations
Highest Quality
  • Rigorous testing to ensure sample integrity and security
Packaging Options
  • Microtiter tubes and caps are available sterile and nonsterile
Specifications
Certifications/ComplianceCertified free of RNase, DNase, Pyrogen, Bioburden, ATP, PCR Inhibitors, Endotoxin and DNA.
Closure MaterialPolyethylene (Plug)
FormatRobotic Rack
Product LineThermo Scientific
Quantity96 tubes/rack 10 racks/unit, 5 units case
ColorNatural
Product TypeMicrotiter Tube
SterilitySterile
Volume (Metric)1.2 mL
Unit SizeCase of 4800