Microtiter Tubes and Racks
Thermo Scientific™
Microtiter Tubes and Racks
Seal microtiter tubes tightly for storage with these polyethylene plug strips -- for use with 1.2mL MCT titertube racks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeSterilityFormatUnit Size
3487PK
also known as 3487
Microtiter TubeSterileRobotic RackPack of 960
3496PK
also known as 3496
Microtiter TubeNon-sterileRobotic RackPack of 960
3492PK
also known as 3492
Microtiter TubeNon-sterileBulkPack of 1000
3487Microtiter TubeSterileRobotic RackCase of 4800
3492Microtiter TubeNon-sterileBulkCase of 10000
Catalog number 3487PK
also known as 3487
Price (USD)
64.29
Online exclusive
79.60
Save 15.31
Pack of 960
Product Type:
Microtiter Tube
Sterility:
Sterile
Format:
Robotic Rack
Unit Size:
Pack of 960
Price (USD)
64.29
Online exclusive
79.60
Save 15.31
Pack of 960
Seal microtiter tubes tightly for storage with polyethylene Thermo Scientific™ Microtiter Tube Plug Strips — for use with 1.2mL MCT Titertube Racks. The 1.2mL microtiter tubes offer additional volume capacity for sample dilution. These are available in a pre-racked 8 x 12 format.
Microtiter Tubes and Caps
  • Polyethylene plug strip seals the microtiter tubes tightly for storage
  • Eight-plug strips can be easily cut apart for individual samples
  • 1.2mL microtiter tubes have additional volume capacity for sample dilution and are ideal for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control and blood bank sample freezer storage
  • Frosted writing areas on each tube and an alphanumeric rack simplify organization and identification of specific samples
  • Microtiter tube system is available in bulk packaging or with a SBS standard footprint
  • Robotic compatible rack can easily integrated into automated liquid handling workstations
Highest Quality
  • Rigorous testing to ensure sample integrity and security
Packaging Options
  • Microtiter tubes and caps are available sterile and nonsterile
Specifications
Certifications/ComplianceCertified free of RNase, DNase, Pyrogen, Bioburden, ATP, PCR Inhibitors, Endotoxin and DNA.
Closure MaterialPolyethylene (Plug)
FormatRobotic Rack
Product LineThermo Scientific
Quantity96 tubes/rack 10 racks/unit, 5 units case
ColorNatural
Product TypeMicrotiter Tube
SterilitySterile
Volume (Metric)1.2 mL
Unit SizePack of 960