2,5-Dimethyl-2,5-di(tert-butylperoxy)hexane, 92%, Thermo Scientific Chemicals
2,5-Dimethyl-2,5-di(tert-butylperoxy)hexane, 92%, Thermo Scientific Chemicals
2,5-Dimethyl-2,5-di(tert-butylperoxy)hexane, 92%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,5-Dimethyl-2,5-di(tert-butylperoxy)hexane, 92%, Thermo Scientific Chemicals

CAS: 78-63-7 | Molecular Formula: C16H34O4 | Molecular Weight: 290.44 g/mol
Have Questions?
Catalog NumberQuantityPackaging
349901000100 gPlastic bottle
Catalog number 349901000
Price (USD)
65.40
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
65.40
Each
Chemical Identifiers
CAS78-63-7
IUPAC Name2,5-bis(tert-butylperoxy)-2,5-dimethylhexane
Molecular FormulaC16H34O4
InChI KeyDMWVYCCGCQPJEA-UHFFFAOYSA-N
SMILESCC(C)(C)OOC(C)(C)CCC(C)(C)OOC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Hydroperoxides=<0.3 % (as 2,5-Dihydroperoxy-2,5-dimethyl-hexane)
Hydroperoxides=<0.3 % (as 2,5-Dihydroperoxy-2,5-dimethyl-hexane)

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators

RUO – Research Use Only