Drierite™, with indicator, 10-20 mesh
Drierite™, with indicator, 10-20 mesh
Drierite™, with indicator, 10-20 mesh
Thermo Scientific Chemicals

Drierite™, with indicator, 10-20 mesh

CAS: 7778-18-9 | CaO4S | 136.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
10 kg
Packaging:
View all
Glass Bottle
Catalog number 350010020
Price (USD)
224.65
Special offer
Online exclusive
Ends: 28-Dec-2026
264.00
Save 39.35
Each
Quantity:
2 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
224.65
Special offer
Online exclusive
Ends: 28-Dec-2026
264.00
Save 39.35
Each
Chemical Identifiers
CAS7778-18-9
IUPAC Namecalcium sulfate
Molecular FormulaCaO4S
InChI KeyOSGAYBCDTDRGGQ-UHFFFAOYSA-L
SMILES[Ca++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue
Appearance (Form)Irregular granules
Water=<0.3 %
Composition>=98 % (CaSO4) Typical
Composition=<2 % (CoCl2) Typical
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, Indicator
RUO – Research Use Only