chloro(diethyl)phosphine, 95%
chloro(diethyl)phosphine, 95%
chloro(diethyl)phosphine, 95%
Thermo Scientific Chemicals

chloro(diethyl)phosphine, 95%

CAS: 686-69-1 | C4H10ClP | 124.55 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3503100101 gGlass bottle
Catalog number 350310010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS686-69-1
IUPAC Namechlorodiethylphosphane
Molecular FormulaC4H10ClP
InChI KeyINJBDKCHQWVDGT-UHFFFAOYSA-N
SMILESCCP(Cl)CC
View more
SpecificationsSpecification SheetSpecification Sheet
NMR>=94 %
Infrared spectrumConforms
Refractive index1.4740 to 1.4760 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only