Barium Peroxide, 95%
Barium Peroxide, 95%
Barium Peroxide, 95%
Thermo Scientific Chemicals

Barium Peroxide, 95%

CAS: 1304-29-6 | BaO2 | 169.33 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3504100252.5 kgPlastic bottle
Catalog number 350410025
Price (USD)
379.00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
379.00
Each
Chemical Identifiers
CAS1304-29-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Beige
Titration Iodometric>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only