4-Bromobenzylamine, 97+%
4-Bromobenzylamine, 97+%
4-Bromobenzylamine, 97+%
4-Bromobenzylamine, 97+%
Thermo Scientific Chemicals

4-Bromobenzylamine, 97+%

CAS: 3959-07-7 | C7H8BrN | 186.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 350530250
Price (USD)
783.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
783.00
Each
Chemical Identifiers
CAS3959-07-7
IUPAC Name1-(4-bromophenyl)methanamine
Molecular FormulaC7H8BrN
InChI KeyXRNVSPDQTPVECU-UHFFFAOYSA-N
SMILESNCC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
GC>=97.0 %
Appearance (Form)Liquid or solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only