Pierce™ Fast Western Kit, SuperSignal™ West Pico, Mouse
Thermo Scientific™

Pierce™ Fast Western Kit, SuperSignal™ West Pico, Mouse

Thermo Scientific Pierce Fast Western Blot Kit, SuperSignal West Pico Substrate contains optimized reagents and protocol for picogram-level detection andRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
35060800 mL
35065200 mL
Catalog number 35060
Price (USD)
301.65
Online exclusive
359.00
Save 57.35
Each
Quantity:
800 mL
Request bulk or custom format
Price (USD)
301.65
Online exclusive
359.00
Save 57.35
Each

Thermo Scientific Pierce Fast Western Blot Kit, SuperSignal West Pico Substrate contains optimized reagents and protocol for picogram-level detection and and Western blot development less than 1 hour after gel transfer.

Features of this kit:

  • Fast—picogram levels of detection in about 1 hour
  • Convenient—no expensive hardware or vacuum required; no clogging issues
  • Simple—optimized protocol makes Western blot analysis easier than ever
  • Economical—cost as little as $14 per blot; no expensive consumables or extra equipment
  • Excellent stability—kit is stable for 1 year stored at 4°C
  • Easy—complete kits contain all components needed to block, probe and develop a blot with your mouse primary antibody

These Fast Western Blot Kits require minimal hands-on time and yield results comparable to classic Western blotting with SuperSignal West Pico Chemiluminescent Substrate.The kits contain all the reagents necessary to complete a Western blot being probed with either a mouse or rabbit primary antibody. Like other Western blotting methods, the included SuperSignal West Pico Substrate produces a signal that is detectable using photographic or other imaging methods. The developed blots can be repeatedly exposed and stripped of the primary antibody and reprobed.

The Pierce Fast Western Blot Kits are reagent-based system that provide optimized reagents for blocking, antibody dilution and detection of Western blots. The Fast Western Blot Kit protocol is quick and efficient method for developing Western blots without the need to purchase expensive instrumentation and consumables required by other popular rapid Western blotting systems.

Related Products

  • Pierce™ Fast Western Kit, SuperSignal™ West Pico, Rabbit (Cat. No. 35066)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ConjugateHRP
ReactivityMouse
SensitivityPicogram-level
Sufficient For20 Mini Blots
Detection MethodChemiluminescence
For Use With (Application)Western Blot
FormLiquid
Membrane CompatibilityNitrocellulose, PVDF
Product LinePierce, SuperSignal
Product TypeWestern Blot Kit
Quantity800 mL
SubstrateHorseradish Peroxidase
Unit SizeEach
Contents & Storage
Sufficient For: 20 mini blots probed with mouse primary antibodies
• Fast Western Antibody Diluent, 400 mL
• Fast Western 10X Wash Buffer, 200 mL
• Fast Western Mouse Optimized HRP Reagent, Pico, 20 mL
• SuperSignal West Pico Luminol/Enhancer Solution, 100 mL
• SuperSignal West Stable Peroxide Solution, 100 mL

Store kit at 4°C.

Frequently asked questions (FAQs)

How can I increase the sensitivity of my Western blot?

Consider transferring to a different membrane or using a different detection method. We have observed increased sensitivity when using PVDF membranes in place of nitrocellulose. On PVDF membranes, using as little as 1 µg of total rat brain protein, PKC can be detected with alkaline phosphatase-mediated chromogenic detection in some cases using affinity-purified antibodies at a concentration of 0.5 µg/mL. Detection sensitivity can also be increased by using chemiluminescent detection, especially when using a SuperSignal West enhanced chemiluminescence subtrate (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/western-blotting/detect-proteins-western-blot/western-blot-detection-reagents/detection-technologies-western-blotting/chemiluminescent-western-blot-detection/supersignal-chemiluminescent-substrates.html) such as SuperSignal West Pico PLUS, SuperSignal West Dura, or SuperSignal West Femto. The secondary antibody should be used as recommend by the manufacturer and optimized as needed.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.