Pierce™ Fast Western Blot Kits, SuperSignal™ West Femto, Mouse
Thermo Scientific™

Pierce™ Fast Western Blot Kits, SuperSignal™ West Femto, Mouse

The Thermo Scientific Pierce Fast Western Blot Kit, SuperSignal West Femto Substrate contains optimized reagents and protocol for mid-to-low femtogram-levelRead more
Have Questions?
Catalog NumberQuantity
35080400 mL
Catalog number 35080
Price (USD)
626.00
Each
Quantity:
400 mL
Request bulk or custom format
Price (USD)
626.00
Each
The Thermo Scientific Pierce Fast Western Blot Kit, SuperSignal West Femto Substrate contains optimized reagents and protocol for mid-to-low femtogram-level detection and typical Western blot development less than 1 hour after gel transfer.

Features of this kit:

• Fast—femtogram levels of detection in about 1 hour
• Convenient—no expensive hardware or vacuum required; no clogging issues
• Simple—optimized protocol makes Western blot analysis easier than ever
• Easy—complete kits contain all components needed to block, probe and develop a blot with your mouse primary antibody

The Fast Western Blot Kit protocol is a quick and efficient method for developing Western blots without the need to purchase expensive instrumentation and consumables required by other rapid Western blotting systems. These complete Pierce Fast Western Blot Kits require minimal hands-on time and yield results comparable to classic Western blotting with SuperSignal West Femto Chemiluminescent Substrate. The kits include SuperSignal West Femto Substrate, the most sensitive enhanced chemiluminescent substrate available for detecting horseradish peroxidase (HRP). This substrate's sensitivity, intensity and duration of the signal allow for easy documentation using photographic or other imaging methods. The developed blots can be repeatedly exposed and stripped of the primary antibody and reprobed. The complete kit contains all the reagents necessary to block and probe a Western blot with a mouse primary antibody.

Related Products
Pierce™ Fast Western Blot Kits, SuperSignal™ West Femto, Rabbit
WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ConjugateHRP
DescriptionPierce Fast Western Blot Kit, SuperSignal West Femto Substrate, Mouse
ReactivityMouse
SensitivityMid-to-low femtogram
Sufficient For10 western blots (800 sq. cm of membrane)
Detection MethodChemiluminescence
For Use With (Application)Western Blot
FormLiquid
Membrane CompatibilityNitrocellulose, PVDF
Product LinePierce, SuperSignal
Product TypeWestern Blot Kit
Quantity400 mL
SubstrateHorseradish Peroxidase
Unit SizeEach
Contents & Storage
Sufficient For: 10 mini blots probed with mouse primary antibodies
• Fast Western Antibody Diluent, 200 mL
• Fast Western 10X Wash Buffer, 100 mL
• Fast Western Mouse Optimized HRP Reagent, Femto, 10 mL
• SuperSignal West Femto Luminol/Enhancer Solution, 50 mL
• SuperSignal West Stable Peroxide Solution, 50 mL

Store kit at 4°C.

Frequently asked questions (FAQs)

How can I increase the sensitivity of my Western blot?

Consider transferring to a different membrane or using a different detection method. We have observed increased sensitivity when using PVDF membranes in place of nitrocellulose. On PVDF membranes, using as little as 1 µg of total rat brain protein, PKC can be detected with alkaline phosphatase-mediated chromogenic detection in some cases using affinity-purified antibodies at a concentration of 0.5 µg/mL. Detection sensitivity can also be increased by using chemiluminescent detection, especially when using a SuperSignal West enhanced chemiluminescence subtrate (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/western-blotting/detect-proteins-western-blot/western-blot-detection-reagents/detection-technologies-western-blotting/chemiluminescent-western-blot-detection/supersignal-chemiluminescent-substrates.html) such as SuperSignal West Pico PLUS, SuperSignal West Dura, or SuperSignal West Femto. The secondary antibody should be used as recommend by the manufacturer and optimized as needed.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.