Methyl 2,6-dichlorophenylacetate, 99%
Methyl 2,6-dichlorophenylacetate, 99%
Methyl 2,6-dichlorophenylacetate, 99%
Thermo Scientific Chemicals

Methyl 2,6-dichlorophenylacetate, 99%

CAS: 54551-83-6 | C9H8Cl2O2 | 219.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 351070250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS54551-83-6
IUPAC Namemethyl 2-(2,6-dichlorophenyl)acetate
Molecular FormulaC9H8Cl2O2
InChI KeyFCWRUYPZZJPCCG-UHFFFAOYSA-N
SMILESCOC(=O)CC1=C(Cl)C=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.5400 to 1.5420 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only